Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4 Mouse anti-Goat, Hamster, Human, Human, Monkey, Mouse, Non-human Primate, Rat, Clone: nan, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA537825

Encompass

Catalog No. PIMA537825


Add to cart

Description

Description

Positive Samples: LO2, U-87MG, MCF7, K-562, Jurkat, Mouse skeletal muscle, Mouse kidney, Mouse heart, Rat brain

COX4 Monoclonal antibody specifically detects COX4 in Goat, Hamster, Human, Human, Monkey, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot
Specifications

Specifications

COX4
Monoclonal
Unconjugated
COX4I1
Liquid
Mouse
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12857, 1327, 29445, 84682, 84683, 84701
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10888, P13073, P19783, Q91W29, Q91Y94, Q96KJ9
AL024441; COX; cox iv; COX IV-1; Cox IV-2; cox4; COX4-1; Cox4-2; Cox4a; Cox4b; cox4i1; cox4i1.S; Cox4i2; COX4L2; COXIV; COXIV-1; CoxIV-2; Cytochrome c oxidase polypeptide IV; cytochrome c oxidase subunit 4 isoform 1, mitochondrial; Cytochrome c oxidase subunit 4 isoform 2, mitochondrial; cytochrome c oxidase subunit 4I1; cytochrome c oxidase subunit 4I2; cytochrome c oxidase subunit IV; cytochrome c oxidase subunit IV isoform 1; cytochrome c oxidase subunit IV isoform 1 precursor; cytochrome c oxidase subunit IV isoform 1 S homeolog; cytochrome c oxidase subunit IV isoform 2; cytochrome c oxidase subunit IV isoform 2 (lung); cytochrome c oxidase subunit IV precursor EC 1.9.3.1; cytochrome c oxidase subunit IV-like 2; cytochrome c oxidase, subunit 4a; cytochrome c oxidase, subunit 4b; cytochrome c oxidase, subunit IV; cytochrome c oxidase, subunit IVa; cytochrome c oxidase, subunit IVb; cytochrome c oxydase subunit 4; dJ857M17.2; FLJ23483; hypothetical protein MGC73355; IV-1; mg:bb02d03; MGC72016; XELAEV_18024950mg; zgc:110058; zgc:73355
A synthetic peptide of human COX IV
Affinity chromatography
RUO
Antibody
Monoclonal
Goat, Hamster, Human, Human, Monkey, Mouse, Non-human Primate, Rat
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit