Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4 Mouse anti-Human, Mouse, Rat, DyLight 488, Clone: 854, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243540G

Catalog No. NBP243540G


Add to cart

Description

Description

COX4 Monoclonal antibody specifically detects COX4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

COX4
854
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
COX4I1
Recombinant protein encompassing a sequence within the center region of human COX4. The exact sequence is proprietary.
Protein G purified
Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Loading Controls, Neuroendocrinology
Primary
1327
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016
Mouse
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit