Learn More
Abnova™ COX4I1 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00001327T01
Catalog No. 89-940-273
Add to cart
Description
Description
Specifications
Specifications
Western Blot
COX4I1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 18.7kDa
Transient overexpression cell lysate was tested with anti-COX4I1 antibody (Mfr. No. H00001327-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_001861.2
pCMV-COX4I1 full-length
100 μL
Human