Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4NB Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549300

Catalog No. PA549300

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

Specifications

Specifications

COX4NB
Polyclonal
PBS with 0.09% sodium azide
O43402
ER membrane protein complex subunit 8, Neighbor of COX4, Protein FAM158B, EMC8, C16orf2, C16orf4, COX4AL, COX4NB, FAM158B, NOC4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10328
Western Blot
Unconjugated
EMC8
Liquid
EMC8
KLH conjugated synthetic peptide between 78-106 amino acids from the Central region of human COX4NB.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit