Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX4NB Mouse anti-Human, Clone: OTI2A4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526318

Catalog No. PIMA526318


Add to cart

Description

Description

Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer and proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit IV isoform 1 of the human mitochondrial respiratory chain enzyme. It is located at the 3 ft. of the NOC4 (neighbor of COX4) gene in a head-to-head orientation, and shares a promoter with it.
Specifications

Specifications

COX4NB
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43402
ER membrane protein complex subunit 8, Neighbor of COX4, Protein FAM158B, EMC8, C16orf2, C16orf4, COX4AL, COX4NB, FAM158B, NOC4
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10328
Immunohistochemistry (Paraffin), Western Blot
OTI2A4
Unconjugated
EMC8
Liquid
EMC8
Full length human recombit protein of COX4NB produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit