Learn More
COX6A1 Antibody (OTI4G2), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270427H
Catalog No. NBP270427H
Add to Cart
Description
Description
COX6A1 Monoclonal specifically detects COX6A1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
COX6A1
Monoclonal
HRP
PBS with No Preservative
COX6A1
Full length recombinant protein of human COX6A1 (NP_004364) produced in HEK293T cells.
0.1 mL
1337.0
Store at 4°C in the dark.
Western Blot
OTI4G2
Western Blot
COX VIa-L, COX6A, COX6AL, Cytochrome c oxidase polypeptide VIa-liver, cytochrome c oxidase subunit 6A1, mitochondrial, cytochrome C oxidase subunit VIa homolog, cytochrome c oxidase subunit VIa polypeptide 1, Cytochrome c oxidase subunit VIA-liver, MGC104500
Mouse
Affinity Purified
Primary
Human
IgG1
Product Suggestions
Product Suggestions