Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
COX6A1 Antibody (OTI4G2), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270427JF646
Description
COX6A1 Monoclonal specifically detects COX6A1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
COX6A1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
COX6A1 | |
Full length recombinant protein of human COX6A1 (NP_004364) produced in HEK293T cells. | |
0.1 mL | |
1337.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4G2 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
COX VIa-L, COX6A, COX6AL, Cytochrome c oxidase polypeptide VIa-liver, cytochrome c oxidase subunit 6A1, mitochondrial, cytochrome C oxidase subunit VIa homolog, cytochrome c oxidase subunit VIa polypeptide 1, Cytochrome c oxidase subunit VIA-liver, MGC104500 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction