Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

COX7C Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551284

Catalog No. PA551284


Add to cart

Description

Description

The antibody detects endogenous levels of total COX7C protein.

Cytochrome c oxidase (COX), the terminal component of the mitochondrial respiratory chain, catalyzes the electron transfer from reduced cytochrome c to oxygen. This component is a heteromeric complex consisting of 3 catalytic subunits encoded by mitochondrial genes and multiple structural subunits encoded by nuclear genes. The mitochondrially-encoded subunits function in electron transfer, and the nuclear-encoded subunits may function in the regulation and assembly of the complex.
Specifications

Specifications

COX7C
Polyclonal
Unconjugated
COX7C
Liquid
COX7C
Synthetic peptide of human COX7C
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
2.5 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P15954, P17665, P80432
COX7C, Cytochrome c oxidase polypeptide VIIc, Cytochrome c oxidase subunit 7C, mitochondrial, cytochrome c oxidase subunit VIIc
Rabbit
IgG
100 μL
-20°C
Primary
100188937, 12867, 1350
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit