Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Coxsackievirus A6 VP2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP313094

Catalog No. NB176781


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Coxsackievirus A6 VP2 Polyclonal antibody specifically detects Coxsackievirus A6 VP2 in Virus samples. It is validated for Western Blot
Specifications

Specifications

Coxsackievirus A6 VP2
Polyclonal
PBS, 20% Glycerol (pH7)
Rabbit
Antigen Affinity-purified
RUO
Virus
Purified
Western Blot
Unconjugated
Capsid protein VP2, CVA6 VP2, P1B, Viral protein 2, Virion protein 2
Recombinant protein encompassing a sequence within the center region of Coxsackievirus A6 VP2. The exact sequence is proprietary.
100 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.