Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP 100356 hydrochloride, Tocris Bioscience™

Catalog Number p-4816996
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
142715-48-8
C31H37ClN4O6
597.109
WWCHXVYTCMPAMV-UHFFFAOYSA-N
cp 100356 hydrochloride, cp-100356 hydrochloride, cp-100356 monohydrochloride, 4-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxy-2-quinazolinamine monohydrochloride, cp-100356 monohydrochloride hplc, 4-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl-n-2 2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxy-2-quinazolinamine hydrochloride, 4-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxyquinazolin-2-amine hydrochloride, 4-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxyquinazolin-2-amine-hydrogen chloride 1/1
71312025
4-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine;hydrochloride
COC1=C(C=C(C=C1)CCNC2=NC3=CC(=C(C=C3C(=N2)N4CCC5=CC(=C(C=C5C4)OC)OC)OC)OC)OC.Cl
This item is not returnable. View return policy
142715-48-8
C31H37ClN4O6
597.109
WWCHXVYTCMPAMV-UHFFFAOYSA-N
cp 100356 hydrochloride, cp-100356 hydrochloride, cp-100356 monohydrochloride, 4-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxy-2-quinazolinamine monohydrochloride, cp-100356 monohydrochloride hplc, 4-3,4-dihydro-6,7-dimethoxy-2 1h-isoquinolinyl-n-2 2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxy-2-quinazolinamine hydrochloride, 4-6,7-dimethoxy-3,4-dihydro-1h-isoquinolin-2-yl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxyquinazolin-2-amine hydrochloride, 4-6,7-dimethoxy-3,4-dihydroisoquinolin-2 1h-yl-n-2-3,4-dimethoxyphenyl ethyl-6,7-dimethoxyquinazolin-2-amine-hydrogen chloride 1/1
71312025
4-(6,7-dimethoxy-3,4-dihydro-1H-isoquinolin-2-yl)-N-[2-(3,4-dimethoxyphenyl)ethyl]-6,7-dimethoxyquinazolin-2-amine;hydrochloride
COC1=C(C=C(C=C1)CCNC2=NC3=CC(=C(C=C3C(=N2)N4CCC5=CC(=C(C=C5C4)OC)OC)OC)OC)OC.Cl

P-gp inhibitor

High affinity P-glycoprotein (P-gp) inhibitor (Ki values are 58 and 94 nM for mouse Pgp1a and Pgp1b isoforms). Inhibits calcein-AM uptake in MDR1-transfected MDCKII cells (IC50 = 0.5 μM) and prazosin transport in BCRP-transfected MDCKII cells (IC50 = 1.5 μM). Displays weak or no inhibitory activity against MRP1, OATP1B1 and several major human P450 enzymes (IC50 > 50 μM).
Quantity 10 mg
Formula Weight 597.1
Percent Purity >98%
Chemical Name or Material CP 100356 hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit