Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP 31398 dihydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1217195-61-3
C22H29ClN4O2
416.95
HUVADZBUMSMSIQ-IVKCLRODSA-N
cp-31398 dihydrochloride hydrate
131698900
N-[2-[(E)-2-(4-methoxyphenyl)ethenyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine;hydrate;hydrochloride
CN(C)CCCNC1=NC(=NC2=CC=CC=C21)C=CC3=CC=C(C=C3)OC.O.Cl
This item is not returnable. View return policy
1217195-61-3
C22H29ClN4O2
416.95
HUVADZBUMSMSIQ-IVKCLRODSA-N
cp-31398 dihydrochloride hydrate
131698900
N-[2-[(E)-2-(4-methoxyphenyl)ethenyl]quinazolin-4-yl]-N',N'-dimethylpropane-1,3-diamine;hydrate;hydrochloride
CN(C)CCCNC1=NC(=NC2=CC=CC=C21)C=CC3=CC=C(C=C3)OC.O.Cl

p53-stabilizing agent

p53 stabilizing agent. Stabilizes the active conformation of p53 and promotes p53 activity in cancer cell lines with mutant or wild-type p53. Inhibits growth of small human tumor xenografts in vivo.
Quantity 50mg
Formula Weight 435.39
Percent Purity >99%
Chemical Name or Material CP 31398 dihydrochloride

Recommended Storage : Desiccate at Room Temperature