Learn More
CP 339818 hydrochloride, Tocris
Non-peptide, potent KV1.3 channel blocker
Manufacturer:  R&D Systems 1399/10
Quantity 10mg
 View more versions of this product
Catalog No. 13-991-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
340.89
CP 339818 hydrochloride
C21H24N2·HCl
N-[1-(Phenylmethyl)-4(1H)-quinolinylidene]-1 pentanamine hydrochloride
Specifications
10mg
>99%
340.89
CP 339818 hydrochloride
478341-55-8
C21H24N2·HCl
N-[1-(Phenylmethyl)-4(1H)-quinolinylidene]-1 pentanamine hydrochloride
Safety and Handling
Recommended Storage : Store at Room Temperature