Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CP 339818 hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent, non-peptide KV1.3 channel antagonist that preferentially binds to the C-type inactivated state of the channel (IC50 ∽ 200 nM) . Inhibits KV1.4 with an IC50 of ∽ 300 nM. Selective over KV1.1, KV1.2, KV1.5, KV1.6, KV3.1-4, and KV4.2.
Specifications
Quantity | 50 mg |
Formula Weight | 340.89 |
Percent Purity | >99% |
Chemical Name or Material | CP 339818 hydrochloride |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction