Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CP 376395 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Description
CP 376395 hydrochloride is a potent and selective CRF1 receptor antagonist (Ki values are 12 and >10000 nM for CRF1 and CRF2 receptors respectively). Binds at an allosteric site. Attenuates CRF-induced activation of the HPA axis in vivo following i.v. administration. Orally active.
Specifications
Specifications
Quantity | 10 mg |
Product Type | CP 376395 hydrochloride |
Purity | >99% |
Molecular Weight (g/mol) | 362.94 |
Inhibitors | CP 376395 hydrochloride |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction