Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CP 724714
Potent and selective ErbB2 inhibitor
Supplier: Tocris Bioscience™ 5973/50
This item is not returnable.
View return policy
Description
CP 724714 is a potent and selective ErbB2 inhibitor (IC50 = 10 nM). Exhibits 500-1000-fold selectivity for ErbB2 over ErbB1 and other related RTKs. Induces G1 cell cycle arrest in an ErbB2-overexpressing human cancer cell line. Promotes tumor apoptosis. Induces tumor regression in a human tumor xenograft model. Displays no apparent toxic effect on cardiac tissue.Specifications
2-Methoxy-N-[(2E)-3-[4-[[3-methyl-4-[(6-methyl-3-pyridinyl)oxy]phenyl]amino]-6-quinazolinyl]-2-propen-1-yl]acetamide | |
50 mg | |
C27H27N5O3 | |
>98% |
383432-38-0 | |
EGF Receptor Inhibitors | |
469.53 |
Safety and Handling
Recommended Storage : Store at -20°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title