Learn More
CP 775146, Tocris™
Selective, high affinity PPARα agonist
Manufacturer:  R&D Systems 4190/50
Quantity 50mg
 View more versions of this product
Catalog No. 41-905-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
423.54
CP 775146
C26H33NO4
2-Methyl-2-[3-[(3S)-1-[2-[4-(1-methylethyl)phenyl]acetyl]-3-piperidinyl]phenoxy]-propanoic acid
Specifications
702680-17-9
C26H33NO4
2-Methyl-2-[3-[(3S)-1-[2-[4-(1-methylethyl)phenyl]acetyl]-3-piperidinyl]phenoxy]-propanoic acid
CP 775146
>99%
423.54
50mg
Safety and Handling
Recommended Storage : Store at Room Temperature