Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CP 80633, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
135637-46-6
C18H24N2O3
316.401
LITNEAPWQHVPOK-NORZTCDRSA-N
unii-8si21e44gn component
14010192
5-[3-[[(1S,3R,4R)-3-bicyclo[2.2.1]heptanyl]oxy]-4-methoxyphenyl]-1,3-diazinan-2-one
COC1=C(C=C(C=C1)C2CNC(=O)NC2)OC3CC4CCC3C4
This item is not returnable. View return policy
135637-46-6
C18H24N2O3
316.401
LITNEAPWQHVPOK-NORZTCDRSA-N
unii-8si21e44gn component
14010192
5-[3-[[(1S,3R,4R)-3-bicyclo[2.2.1]heptanyl]oxy]-4-methoxyphenyl]-1,3-diazinan-2-one
COC1=C(C=C(C=C1)C2CNC(=O)NC2)OC3CC4CCC3C4

Selective PDE4 inhibitor

Selective inhibitor of phosphodiesterase type 4 (IC 50 values are 1.9, > 100, > 100, > 100 and > 100 μM for human lung PDE4, lung PDE1, lung PDE2, heart PDE3 and platelet PDE-V respectively) that displays no significant PDE4 isozyme selectivity. Inhibits hydrolysis of cAMP in isolated human peripheral blood monocytes, eosinophils and T cells. Displays anti-inflammatory and bronchodilatory effects in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 316.39
Percent Purity >99%
Chemical Name or Material CP 80633

Recommended Storage : Store at +4°C