Learn More
CPA1 (Carboxypeptidase A1) Mouse anti-Human, Clone: OTI1C12, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA504524
Catalog No. 50-167-5189
Add to cart
Description
Description
CPA1, also known as CPA, is a 419 amino acid secreted monomeric protein that is highly expressed in pancreatic tissue. Functioning as a pancreatic exopeptidase, CPA1 uses zinc as a cofactor to catalyze the release of C-terminal amino acids from a variety of proteins, thereby playing a key role in protein digestion and degradation. Via its catalytic activity, CPA1 is also thought to be involved in zymogen (proenzyme) inhibition, probably functioning to block enzyme activation pathways. Abnormal levels of CPA1 are associated with pancreatic cancer, suggesting a possible role in either tumor progression or tumor suppression events.
Specifications
Specifications
CPA1 (Carboxypeptidase A1)
Monoclonal
0.82 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P15085
CPA
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1357
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1C12
Unconjugated
CPA1
Liquid
CPA1
Full length human recombit protein of human CPA1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human