Learn More
CPA2 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer:  Abnova Corporation H00001358T01
Catalog No. 89-008-816
Add to cart
Description
Description
Western Blotting
Specifications
Specifications
293T
BC014571.1
6.8
Transient overexpression cell Lysate was tested with Anti-CPA2 antibody (H00001358-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
CPA2 293T Cell Transient Overexpression Lysate(Denatured)
45.98
pCMV-CPA2 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot