Learn More
CPA2 Mouse anti-Human, Mouse, Rat, Clone: OTI3D8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF504696

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Three different forms of human pancreatic procarboxypeptidase A have been isolated. The encoded protein represents the A2 form, which is a monomeric protein with different biochemical properties from the A1 and A3 forms. The A2 form of pancreatic procarboxypeptidase acts on aromatic C-terminal residues and is a secreted protein.Specifications
CPA2 | |
Monoclonal | |
Unconjugated | |
CPA2 | |
carboxypeptidase A2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
1358, 232680, 296959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P19222, P48052, Q504N0 | |
CPA2 | |
Full length human recombit protein of human CPA2 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.