Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPC™ Ferruleless Polytube Inserts with Shut-Off Valve

For use in laboratory equipment, medical and surgical instrumentation, water transfer, air systems and other applications

Manufacturer:  CPC™ PMCD2004

 View more versions of this product

Catalog No. 05-112-66


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

6.4mm
For use in laboratory equipment, medical and surgical instrumentation, water transfer, air systems and other applications
White
14.7mm
1.3 to 1.8 in.
Buna-N
-40° to 180°F
Acetal
Shut-off valves
0.58 in.
PTF Valved In-Line Coupling Insert
33 to 45mm
120psi
-40° to 82°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit