Learn More
CPC™ Plastic Quick-Disconnect Fittings
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.
Supplier: CPC™ MPCK17004T39
Description
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables
CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.
- Material: Medical-grade ABS, polycarbonate and polysulfone
- Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
- Fow Rate: Cv∽5.5 (79.25) max.
- Ergonomic thumb latch
- Audible 'click' when connected
- USP Class VI materials
- Can be sterilized by autoclave, EtO, e-beam, or gamma
- Coupling halves can be rotated when connected
- Tapered barb
- BSE/TSE free materials
- Cell culture media packaging and transfer lines
- Harvest and collection applications
- Buffer solutions
- Tube sets
- Urology drainage/collection systems
- Hypo/hyperthermia blankets
Specifications
USP Class VI, ADCF | |
Autoclavable | |
Polysulfone | |
33 mm | |
Cell culture media packaging and transfer lines, harvest and collection applications | |
-40°C | |
0.99 in. | |
Amber | |
300°F |
Hose Barb Non-valved Coupling Body | |
6.4 mm | |
1.3 in. | |
6.4mm | |
-40°F | |
25 mm | |
Lock | |
60 psi | |
149°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.