Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPI17 alpha Mouse anti-Human, DyLight 405, Clone: 4H10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104336V
Description
CPI17 alpha Monoclonal specifically detects CPI17 alpha in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
CPI17 alpha | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PPP1R14A | |
Recombinant human PPP1R14A (1-147aa) purified from E. coli | |
0.1 mL | |
Phospho Specific, Protein Phosphatase | |
94274 | |
Human | |
Purified |
Western Blot, ELISA | |
4H10 | |
Western Blot, ELISA | |
17 kDa PKC-potentiated inhibitory protein of PP1, CPI-1717-kDa PKC-potentiated inhibitory protein of PP1, CPI1717-KDa protein, PKC-potentiated inhibitory protein of PP1, PPP1INL, Protein kinase C-potentiated inhibitor protein of 17 kDa, protein phosphatase 1 regulatory subunit 14A, protein phosphatase 1, regulatory (inhibitor) subunit 14A | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human PPP1R14A mAb, clone 4H10, is derived from hybridization of mouse FO myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PPP1R14A protein. | |
Store at 4C in the dark. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title