Learn More
Abnova™ CPM 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001368T01
Catalog No. 89-940-351
Add to cart
Specifications
Specifications
Western Blot
293T
NM_001005502.1
pCMV-CPM full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
CPM 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 48.84kDa
Transient overexpression cell lysate was tested with anti-CPM antibody (Mfr. No. H00001368-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)