Learn More
CPM Mouse anti-Human, Clone: OTI4E11, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF807364
Catalog No. 50-166-8235
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Specifically removes C-terminal basic residues (Arg or Lys) from peptides and proteins. It is believed to play important roles in the control of peptide hormone and growth factor activity at the cell surface, and in the membrane-localized degradation of extracellular proteins.
Specifications
Specifications
CPM
Monoclonal
Unconjugated
CPM
Lyophilized
CPM
Human recombit protein fragment corresponding to amino acids 124-423 of human CPM produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI4E11
PBS with 8% trehalose and no preservative; pH 7.3
P14384
carboxypeptidase M
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1368