Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CPN1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | CPN1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CPN1 Polyclonal specifically detects CPN1 in Human samples. It is validated for Western Blot.Specifications
CPN1 | |
Polyclonal | |
Rabbit | |
P15169 | |
1369 | |
Synthetic peptides corresponding to CPN1(carboxypeptidase N, polypeptide 1) The peptide sequence was selected from the middle region of CPN1. Peptide sequence EWLGNREALIQFLEQVHQGIKGMVLDENYNNLANAVISVSGINHDVTSGD. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ACBP, Anaphylatoxin inactivator, Arginine carboxypeptidase, Carboxypeptidase N polypeptide 1, carboxypeptidase N polypeptide 1 50 kD, Carboxypeptidase N small subunit, carboxypeptidase N, polypeptide 1, carboxypeptidase N, polypeptide 1, 50kD, CPNcarboxypeptidase N catalytic chain, EC 3.4.17, EC 3.4.17.3, FLJ40792, kininase I, Kininase-1, Lysine carboxypeptidase, Plasma carboxypeptidase B, SCPNcarboxypeptidase N catalytic subunit, Serum carboxypeptidase N | |
CPN1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title