Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPN2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552094
Description
Immunogen sequence: FDTNYNLFNL ALHGNPWQCD CHLAYLFNWL QQYTDRLLNI QTYCAGPAYL KGQVVPALNE KQLVCPVTRD HLGFQVTWPD ESKAGGSWDL AVQERAARSQ CTYSNPEGTV VLACDQAQCR WLNVQLSPWQ GSLGLQYNAS QEW Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 62%.
CPN2, the 83 kDa subunit binds and stabilizes the catalytic subunit at 37 degrees Celsius and keeps it in circulation. Under some circumstances it may be an allosteric modifier of the catalytic subunit.
Specifications
CPN2 | |
Polyclonal | |
Unconjugated | |
CPN2 | |
1300018K11Rik; ACBP; arginine carboxypeptidase; carboxypeptidase N 83 kDa chain; Carboxypeptidase N large subunit; Carboxypeptidase N polypeptide 2; Carboxypeptidase N regulatory subunit; carboxypeptidase N subunit 2; carboxypeptidase N, polypeptide 2; carboxypeptidase N, polypeptide 2 homolog; carboxypeptidase N, polypeptide 2, 83kD; CPN reg; Cpn2; CPNreg; CPN-reg; RGD1305170 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1370 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P22792 | |
CPN2 | |
Recombinant protein corresponding to Human CPN2. Recombinant protein control fragment (Product #RP-100442). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction