Learn More
CPNE6 Mouse anti-Human, Clone: 6.4_1B4, Abnova™
Mouse monoclonal antibody raised against human CPNE6.
Manufacturer:  Abnova Corporation MAB12755
Catalog No. 89-933-991
Add to cart
Description
Description
This gene encodes a brain-specific member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains, and one von Willebrand factor A domain. It may have a role in synaptic plasticity. [provided by RefSeq]
Specifications
Specifications
CPNE6
6.4_1B4
Mouse monoclonal antibody raised against human CPNE6.
In PBS (0.02% sodium azide).
N-COPINE
Mouse
IgG2a
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunocytochemistry (10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
CPNE6
Human HeLa cells
Protein A/G purification
RUO
Primary
9362