Learn More
CPNE7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Thermo Scientific PA598861
Catalog No. PIPA598861
Add to cart
Description
Description
This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
CPNE7
Polyclonal
Unconjugated
CPNE7
Liquid
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
102278, 27132, 361433
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
3.63 mg/mL
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
H1UBN0, Q0VE82, Q9UBL6
Cpne7
Recombit Human Copine-7 protein (254-393AA)
Protein G
RUO
Antibody
Polyclonal
Human, Mouse, Rat