Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPO Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548661

Catalog No. PA548661

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Probable carboxypeptidase which may cleave proteins with C-terminal acidic residues.
Specifications

Specifications

CPO
Polyclonal
PBS with 0.09% sodium azide
Q8IVL8
Carboxypeptidase O, CPO, 3417-, CPO
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
130749
Western Blot
Unconjugated
CPO
Liquid
CPO
KLH conjugated synthetic peptide between 46-75 amino acids from the N-terminal region of human CPO.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit