Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPPED1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559359
Description
Immunogen sequence: MSDAEAGGVF HRARGRTLAA FPAEKESEWK GPFYFILGAD PQFGLIKAWS TGDCDNGGDE WEQEIRLTEQ AVQAINKLNP KPKFFVLCGD LIHAMP Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Protein phosphatase that dephosphorylates AKT family kinase specifically at 'Ser-473', blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
Specifications
CPPED1 | |
Polyclonal | |
Unconjugated | |
CPPED1 | |
C530044N13Rik; calcineurin like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain containing 1; calcineurin-like phosphoesterase domain-containing protein 1; complete S transactivated protein 1; complete S-transactivated protein 1; Cpped1; CSTP1; RGD1306568; Serine/threonine-protein phosphatase CPPED1; Vcalcineurin-like phosphoesterase domain containing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BRF8 | |
CPPED1 | |
Recombinant protein corresponding to Human CPPED1. Recombinant protein control fragment (Product #RP-98656). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction