Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPPED1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548930

Catalog No. PA548930

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protein phosphatase that dephosphorylates AKT family kinase specifically at  ft.Ser-473 ft., blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
Specifications

Specifications

CPPED1
Polyclonal
PBS with 0.09% sodium azide
Q9BRF8
Serine/threonine-protein phosphatase CPPED1, Calcineurin-like phosphoesterase domain-containing protein 1, Complete S-transactivated protein 1, CPPED1, CSTP1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55313
Western Blot
Unconjugated
CPPED1
Liquid
CPPED1
KLH conjugated synthetic peptide between 211-237 amino acids from the C-terminal region of human CPPED1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit