Learn More
CPPED1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206281AP
Catalog No. 50-173-2998
Add to cart
Description
Description
Protein phosphatase that dephosphorylates AKT family kinase specifically at  ft.Ser-473 ft., blocking cell cycle progression and promoting cell apoptosis. May play an inhibitory role in glucose uptake by adipocytes.
Specifications
Specifications
CPPED1
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BRF8
CPPED1, CSTP1
Rabbit
IgG
150 μL
-20°C
Primary
55313
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CPPED1
Liquid
CPPED1
CPPED1 Fusion Protein Ag14572
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human