Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPPHA, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
693288-97-0
C22H15ClN2O4
406.822
UFOUABRZSDGGAZ-UHFFFAOYSA-N
cppha,n-4-chloro-2-1,3-dioxoisoindolin-2-yl methyl phenyl-2-hydroxybenzamide,n-4-chloro-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl methyl phenyl-2-hydroxybenzamide,n-4-chloro-2-1,3-dioxoisoindol-2-yl methyl phenyl-2-hydroxybenzamide,d07fvz,cppha hplc
9931205
N-[4-chloro-2-[(1,3-dioxoisoindol-2-yl)methyl]phenyl]-2-hydroxybenzamide
C1=CC=C2C(=C1)C(=O)N(C2=O)CC3=C(C=CC(=C3)Cl)NC(=O)C4=CC=CC=C4O
This item is not returnable. View return policy
693288-97-0
C22H15ClN2O4
406.822
UFOUABRZSDGGAZ-UHFFFAOYSA-N
cppha,n-4-chloro-2-1,3-dioxoisoindolin-2-yl methyl phenyl-2-hydroxybenzamide,n-4-chloro-2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl methyl phenyl-2-hydroxybenzamide,n-4-chloro-2-1,3-dioxoisoindol-2-yl methyl phenyl-2-hydroxybenzamide,d07fvz,cppha hplc
9931205
N-[4-chloro-2-[(1,3-dioxoisoindol-2-yl)methyl]phenyl]-2-hydroxybenzamide
C1=CC=C2C(=C1)C(=O)N(C2=O)CC3=C(C=CC(=C3)Cl)NC(=O)C4=CC=CC=C4O

Positive allosteric modulator at mGlu1 and mGlu5

Positive allosteric modulator at mGlu1 and mGlu5; thought to act at a novel allosteric site. Potentiates mGlu5 responses by a mechanism distinct from that of VU 29 (Cat. No. 4458).
Quantity 10mg
Formula Weight 406.82
Percent Purity >98%
Chemical Name or Material CPPHA

Recommended Storage : Store at -20°C