Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CPS1 Rabbit anti-Human, Polyclonal, Abnova™

Rabbit Polyclonal Antibody

$346.00

Specifications

Antigen CPS1
Host Species Rabbit
Target Species Human
Applications Immunohistochemistry (PFA fixed), Western Blot
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-033-213 View Documents Abnova Corporation
PAB29925
100 uL Each for $346.00
Add to cart
 
89-035-695 View Documents Abnova Corporation
PAB30202
40 ug Each for $346.00
Add to cart
 
Description

Description

Rabbit polyclonal antibody raised against synthetic peptide of human CPS1.

  • High specificity
  • High reactivity
Specifications

Specifications

CPS1
Human
RUO
Liquid
Primary
In PBS (2% sucrose, 0.09% sodium azide).
Rabbit
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit