Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CPSF4L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560603
Description
Immunogen sequence: GLERFTFAFE KDVEMQKGTG LLPFQGMDKS ASAVCNFFTK GLCEKGKLCP FRHDR Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.

Specifications
CPSF4L | |
Polyclonal | |
Unconjugated | |
Cpsf4l | |
0610010C04Rik; 1500000C01Rik; cleavage and polyadenylation specific factor 4 like; cleavage and polyadenylation specific factor 4-like; Cpsf4l; D11Ertd636e; putative cleavage and polyadenylation specificity factor subunit 4-like protein; RGD1306397 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
642843 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NMK7 | |
Cpsf4l | |
Recombinant protein corresponding to Human CPSF4L. Recombinant protein control fragment (Product #RP-99971). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction