Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ CPYPP
DOCK2-Rac1 interaction inhibitor
$167.04 - $693.68
Specifications
Chemical Name or Material | Rho GTPase Inhibitors |
---|---|
CAS | 310460-39-0 |
Target | Small G Protein Inhibitors |
Molecular Formula | C18H13ClN2O2 |
Formula Weight | 324.76 |
Description
DOCK2-Rac1 interaction inhibitor; suppresses DOCK2-induced Rac1 activation. Also inhibits DOCK1 and DOCK5. Exhibits selectivity over DOCK9. Reduces chemotactic responses in vitro and in vivo, and activates T cells in vitro. Also attenuates HER2-mediated breast cancer cell migration in vitro.Specifications
Rho GTPase Inhibitors | |
Small G Protein Inhibitors | |
324.76 |
310460-39-0 | |
C18H13ClN2O2 | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title