Learn More
CR2 Mouse anti-Human, Clone: Bu32, Abnova™
Mouse monoclonal antibody raised against human CR2.
Manufacturer:  Abnova Corporation MAB12691
Catalog No. 89-933-998
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Complement component receptor-2 (CR2) is the membrane protein on B lymphocytes to which the Epstein-Barr virus (EBV) binds during infection of these cells. See also CR1 (MIM 120620). Yefenof et al. (1976) [PubMed 181330] found complete overlapping of EBV receptors and C3 (MIM 120700) receptors on human B lymphocytes.[supplied by OMIM]
Specifications
Specifications
CR2
Bu32
Mouse monoclonal antibody raised against human CR2.
In PBS, pH 7.4 (0.09% sodium azide).
C3DR/CD21/SLEB9
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (PFA fixed), Immunoprecipitation
Unconjugated
Flow Cytometry (1:50-1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Liquid
CR2
Human lymphocytes
Protein G purification
RUO
Primary
1380