Learn More
CRABP2 Antibody (OTI10D6), DyLight 680, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270448FR
Catalog No. NBP270448FR
Add to Cart
Description
Description
CRABP2 Monoclonal specifically detects CRABP2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
Specifications
CRABP2
Monoclonal
DyLight 680
PBS with 0.05% Sodium Azide
CRABP2
Full length recombinant protein of human CRABP2 (NP_001869) produced in E.coli.
0.1 mL
Primary
Human, Mouse
IgG1
Western Blot, Flow Cytometry, Immunohistochemistry
OTI10D6
Western Blot, Flow Cytometry, Immunohistochemistry
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6
Mouse
Affinity Purified
Core ESC Like Genes, Neuroscience, Stem Cell Markers
1382.0
Store at 4°C in the dark.
Recommendations
Recommendations