Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRABP2 Mouse anti-Human, Biotin, Clone: OTI10D6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP270448B
Description
CRABP2 Monoclonal antibody specifically detects CRABP2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
CRABP2 | |
OTI10D6 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
CRABP2 | |
Full length recombinant protein of human CRABP2 (NP_001869) produced in E.coli. | |
Immunogen affinity purified | |
Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
Primary | |
1382.0 |
Immunohistochemistry, Western Blot | |
Biotin | |
PBS with 0.05% Sodium Azide | |
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title