Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRABP2 Rabbit anti-Human, DyLight 350, Clone: 16, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289837UV

Catalog No. NB109661


Add to cart

Description

Description

CRABP2 Monoclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

CRABP2
16
50 mM Sodium Borate
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6
This antibody was obtained from a rabbit immunized with purified, recombinant Human CRABP2 (NP_001869.1; Pro2-Glu138).
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 350
Purified
Rabbit
IgG
0.1 mL
Core ESC Like Genes, Neuroscience, Stem Cell Markers
Primary
1382
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit