Sino Biological™ CRABP2 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding human CRABP2 (Pro2-Glu 138) fused to polyhistide tag at N-terminus

Manufacturer: Sino Biological™ 11203H07E5

 View more versions of this product

Catalog No. 11-203-H07E5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 16.5kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Common Name Cellular Retinoic Acid Binding Protein 2
    Gene Symbol CRABP2
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Expression System E.coli
    Product Type CRABP2 Recombinant Human Protein
    Format Lyophilized
    Gene ID (Entrez) 1382

    Expressed from DNA sequence encoding human CRABP2 (Pro2-Glu 138) fused to polyhistide tag at N-terminus

    • Formulation: 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween80, 5% Mannitol, 5% Trehalose, 0.02% Tween80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit