Learn More
CRACC/SLAMF7 Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0200-6G17, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212206AF750
Catalog No. NP212206A70
Add to cart
Description
Description
CRACC/SLAMF7 Monoclonal antibody specifically detects CRACC/SLAMF7 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CRACC/SLAMF7
MM0200-6G17
Flow Cytometry
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
SLAMF7
Human SLAMF7
0.1 mL
Primary
57823