Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRALBP Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI1G6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271275JF646

Catalog No. N271275J646


Add to cart

Description

Description

CRALBP Monoclonal antibody specifically detects CRALBP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications

Specifications

CRALBP
OTI1G6
Flow Cytometry, Immunohistochemistry
Affinity Purified, Conjugated
RLBP1
Full length recombinant protein of human CRALBP (NP_000317) produced in HEK293T cells.
Immunogen affinity purified
Vision
Primary
6017.0
Flow Cytometry, Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit