Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRALBP Mouse anti-Human, Mouse, Rat, PerCP, Clone: OTI1G6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271275PCP
Description
CRALBP Monoclonal specifically detects CRALBP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CRALBP | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
RLBP1 | |
Full length recombinant protein of human CRALBP (NP_000317) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Flow Cytometry | |
OTI1G6 | |
Flow Cytometry | |
Cellular retinaldehyde-binding protein, cellular retinaldehyde-binding protein-1, CRALBPretinaldehyde-binding protein 1, MGC3663, retinaldehyde binding protein 1 | |
Mouse | |
Affinity Purified | |
Vision | |
6017.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title