Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Craniofacial Development Protein 1 Mouse anti-Human, DyLight 650, Clone: 12411, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243760C

Catalog No. NBP243760C


Add to cart

Description

Description

Craniofacial Development Protein 1 Monoclonal antibody specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Craniofacial Development Protein 1
12411
Western Blot
Purified
CFDP1
Recombinant protein encompassing a sequence within the center region of human CFDP1. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti
Mouse
IgG1
0.1 ml
Primary
10428
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit