Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Craniofacial Development Protein 1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259089

Catalog No. NBP259089


Add to cart

Description

Description

Craniofacial Development Protein 1 Polyclonal antibody specifically detects Craniofacial Development Protein 1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

Craniofacial Development Protein 1
Unconjugated
Tris-buffered Saline containing 0.1% BSA with 0.09% Sodium Azide
BCNTBUCENTAUR, Bucentaur, CP27phosphoprotein (Bucentaur), craniofacial development protein 1, p97, SWC5, Yeti
Rabbit
IgG
100 ul
Store at 4C.
Polyclonal
Human
Western Blot, Immunoprecipitation
Western Blot, Immunoprecipitation
Affinity Purified
CFDP1
Maps to a region between 249 and 299
Immunogen affinity purified
RUO
Primary
10428
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only