Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRB3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548875

Catalog No. PA548875

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the establishment of cell polarity in mammalian epithelial cells. Regulates the morphogenesis of tight junctions.
Specifications

Specifications

CRB3
Polyclonal
PBS with 0.09% sodium azide
Q9BUF7, Q8QZT4
Protein crumbs homolog 3, CRB3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
224912, 92359
Western Blot
Unconjugated
CRB3
Liquid
CRB3
KLH conjugated synthetic peptide between 67-95 amino acids from the Central region of human CRB3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit