Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRBN Mouse anti-Human, Unconjugated, Clone: 4D6 (-3H10-D6), MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CRBN
Applications Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABC15931MI
View Documents
MilliporeSigma
MABC1593100UG
100 μg
Each for $372.00
Add to cart
 
MABC15932MI
View Documents
MilliporeSigma
MABC159325UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

CRBN Monoclonal antibody specifically detects CRBN in Human samples. It is validated for Immunoprecipitation, Western Blotting.
Specifications

Specifications

CRBN
Unconjugated
Mouse
Protein Cereblon;CRBN
His-tagged recombinant protein fragment corresponding to the N-terminal region of human Cereblon.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunoprecipitation, Western Blot
Purified
Human
CRBN
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit