Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cre Mouse anti-Cre/lox Systems, Alexa Fluor™ 594, Clone: 10212, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242832AF594

Catalog No. NP242832A94


Add to cart

Description

Description

Cre Monoclonal antibody specifically detects Cre in Cre/lox System samples. It is validated for Western Blot.
Specifications

Specifications

Cre
10212
Western Blot
Cyclization recombinase
The exact sequence is proprietary.
0.1 mL
Primary
Cre/lox Systems
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit